Anti-ZC3HAV1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16820-1-AP-150UL
10097-254EA 462.47 USD
10097-254
Anti-ZC3HAV1 Rabbit Polyclonal Antibody
Antibodies
The zinc-finger antiviral protein (ZAP) was originally identifed from a rat cDNA library due to its conferring resistance to retroviruses. ZC3HAV1 is a CCCH-type zinc finger protein, it may primarily function to inhibit viral gene expression and induce an innate immunity to viral infection. Alternative splicing occurs at this locus and two variants, each encoding distinct isoforms.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ZC3HAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR