Anti-WWOX Rabbit Polyclonal Antibody

Supplier: Proteintech
15299-1-AP-150UL
10097-158EA 462.47 USD
10097-158
Anti-WWOX Rabbit Polyclonal Antibody
Antibodies
WWOX(WW domain-containing oxidoreductase) is also named as FOR, WOX1 and belongs to the short-chain dehydrogenases/reductases (SDR) family. The gene encodes a 46 kDa protein that contains two WW domains and a short-chain dehydrogenase/reductase domain and the protein is lost or reduced in the majority of many cancer types including breast, prostate, esophageal, lung, stomach, and pancreatic carcinomas and in a large fraction of other cancer types. WWOX behaves as a potential tumor-suppressor gene. It has 7 isoforms produced by alternative splicing.

Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000

Type: Primary
Antigen: WWOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR