Anti-WNT6 Rabbit Polyclonal Antibody

Supplier: Proteintech
24201-1-AP-150UL
10097-138EA 462.47 USD
10097-138
Anti-WNT6 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:500-1:5000; IHC: Human small intestine Tissue, 1:20-1:200

Type: Primary
Antigen: WNT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR