Anti-UROC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
25032-1-AP-150UL
10096-812EA 462.47 USD
10096-812
Anti-UROC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Liver Tissue, 1:200-1:1000

Type: Primary
Antigen: UROC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR