Anti-UMPS Rabbit Polyclonal Antibody

Supplier: Proteintech
14830-1-AP-150UL
10096-778EA 462.47 USD
10096-778
Anti-UMPS Rabbit Polyclonal Antibody
Antibodies
UMPS, also named as OPRT and ODC, plays an important role in pyrimidine synthesis, converting orotic acid to uridine 5′ monophosphate. UMPS has four isoforms with MW 53 kDa, 43 kDa, 33 kDa and 23 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: UMPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR