Anti-UBE4B Rabbit Polyclonal Antibody

Supplier: Proteintech
18148-1-AP-150UL
10096-668EA 462.47 USD
10096-668
Anti-UBE4B Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:1000-1:10000; IF: HeLa Cells, 1:10-1:100; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: UBE4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR