Anti-UBE3A Rabbit Polyclonal Antibody

Supplier: Proteintech
10344-1-AP-150UL
10096-664EA 462.47 USD
10096-664
Anti-UBE3A Rabbit Polyclonal Antibody
Antibodies
UBE3A(Ubiquitin-protein ligase E3A) is also named as E6AP, EPVE6AP, HPVE6A and belongs to the E3 ubiquitin protein ligase family. It functions as both an E3 ligase in the ubiquitin proteasome pathway and as a transcriptional coactivator. It is also initially identified as a cellular protein that mediates in vitro association of the human papillomavirus E6 protein with p53, leading to the ubiquitin-dependent degradation of p53. Defects in UBE3A are a cause of Angelman syndrome (AS). It has 3 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: UBE3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR