Anti-UBC13 Rabbit Polyclonal Antibody

Supplier: Proteintech
10243-1-AP-150UL
10096-650EA 462.47 USD
10096-650
Anti-UBC13 Rabbit Polyclonal Antibody
Antibodies
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2N is a member of the E2 ubiquitin-conjugating enzyme family. Studies in mouse suggest that this protein plays a role in DNA postreplication repair

Western Blot: Mouse Thymus Tissue, 1:500-1:5000

Type: Primary
Antigen: UBC13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR