Anti-UBE2G2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10722-1-AP-150UL
10096-638EA 462.47 USD
10096-638
Anti-UBE2G2 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Prostate Cancer Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: UBE2G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR