Anti-UBE2F Rabbit Polyclonal Antibody

Supplier: Proteintech
17056-1-AP-150UL
10096-634EA 462.47 USD
10096-634
Anti-UBE2F Rabbit Polyclonal Antibody
Antibodies
UBE2F(Ubiquitin-conjugating enzyme E2 F) is also named as NCE2(NEDD8-conjugating enzyme 2) and belongs to the ubiquitin-conjugating enzyme family.The canonical uBE2M catalyses the modification of most cullins, whereas uBE2F is specifically responsible for cuL5 neddylation. uBE2F is regulated differently than uBE2M, supporting the idea that neddylation is used to fine-tune ubiquitin-chain formation by different cullin-dependent E3s.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200

Type: Primary
Antigen: UBE2F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR