Anti-UQCRFS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18443-1-AP-150UL
10096-588EA 462.47 USD
10096-588
Anti-UQCRFS1 Rabbit Polyclonal Antibody
Antibodies
The ubiquinol cytochrome c reductase UQCRFS1 is a key subunit of the cytochrome bc1 complex (complex III) of the mitochondrial respiratory chain. The UQCRFS1 gene encode a 30 kDa protein with a transit peptide and it appears to be involved in development of more aggressive phenotype of breast cancer. UQCRFS1 is also named as RISP, complex III subunit 5, cytochrome b-c1 complex subunit 5 and rieske iron-sulfur protein. This antibody is specific to UQCRFS1.

Western Blot: MCF-7 Cells, 1:1000-1:10000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: UQCRFS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR