Anti-TULP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
55155-1-AP-150UL
10096-508EA 462.47 USD
10096-508
Anti-TULP4 Rabbit Polyclonal Antibody
Antibodies
TULP4, also named as KIAA1397, TUBL4 and TUSP, belongs to the TUB family. It may be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. This antibody is specific to TULP4.

Western Blot: Human Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: TULP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR