Anti-TULP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
13637-1-AP-150UL
10096-506EA 462.47 USD
10096-506
Anti-TULP3 Rabbit Polyclonal Antibody
Antibodies
The tubby-like protein 3 (Tulp3) has been identified as a member of tubby and tubby-like proteins (TULPs) that plays an important role in maintenance and function of neuronal cells during development and post-differentiation. Tulp3 gene is essential for embryonic development in mice. Recently TULP3 has been reported to bind to the IFT(intraflagellar transport)-A complex and promote trafficking of G protein-coupled receptors into primary cilia,which is implicated in regulating neuronal function.

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: TULP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR