Anti-TRIM38 Rabbit Polyclonal Antibody

Supplier: Proteintech
13405-1-AP-150UL
10096-344EA 462.47 USD
10096-344
Anti-TRIM38 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: TRIM38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR