Anti-TNPO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20679-1-AP-150UL
10096-276EA 462.47 USD
10096-276
Anti-TNPO1 Rabbit Polyclonal Antibody
Antibodies
TNPO1, also named as KPNB2, MIP1, TRN and MIP, belongs to the importin beta family. TNPO1 functions in nuclear protein import as nuclear transport receptor. It serves as receptor for nuclear localization signals (NLS) in cargo substrates. TNPO1 is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. The directionality of nuclear import is thought to be conferred by an asymmetric distribution of the GTP- and GDP-bound forms of Ran between the cytoplasm and nucleus. TNPO1 is involved in nuclear import of M9-containing proteins. In vitro, it binds directly to the M9 region of the heterogeneous nuclear ribonucleoproteins (hnRNP), A1 and A2 and mediates their nuclear import. It is involved in hnRNP A1/A2 nuclear export. TNPO1 mediates the nuclear import of ribosomal proteins RPL23A, RPS7 and RPL5. It binds to a beta-like import receptor binding (BIB) domain of RPL23A. In vitro, it mediates nuclear import of H2A, H2B, H3 and H4 histones, and SRP19. In case of HIV-1 infection, binds and mediates the nuclear import of HIV-1 Rev. The antibody is specific to TNPO1.

Western Blot: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: TNPO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR