Anti-Transketolase Mouse Monoclonal Antibody [clone: 3E2F3]

Supplier: Proteintech
66016-1-IG-150UL
10096-272EA 462.47 USD
10096-272
Anti-Transketolase Mouse Monoclonal Antibody [clone: 3E2F3]
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: Transketolase
Clonality: Monoclonal
Clone: 3E2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR