Anti-TMEM161A Rabbit Polyclonal Antibody

Supplier: Proteintech
24898-1-AP-150UL
10096-086EA 462.47 USD
10096-086
Anti-TMEM161A Rabbit Polyclonal Antibody
Antibodies
TMEM161A also named as transmembrane protein 161A is a 479 amino-acid protein, which belongs to TMEM161 family. The molecular weight of TMEM161A is 56 kDa. TMEM161A is multi-pass membrane protein and localizes on the cell membrane. It may involve in the negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage and cellular response to oxidative stress.

Western Blot: MCF-7 Cells, 1:500-1:5000

Type: Primary
Antigen: TMEM161A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR