Anti-TMED2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11981-1-AP-150UL
10096-064EA 462.47 USD
10096-064
Anti-TMED2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse pancreas Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TMED2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR