Anti-TIN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11368-1-AP-150UL
10096-022EA 462.47 USD
10096-022
Anti-TIN2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse spleen Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: TIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR