Anti-TIMP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12326-1-AP-150UL
10096-020EA 462.47 USD
10096-020
Anti-TIMP4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:fetal Human Brain Tissue, 1:200-1:2000; IF: MCF-7 Cells, 1:10-1:100; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: TIMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR