Anti-tim Rabbit Polyclonal Antibody

Supplier: Proteintech
14421-1-AP-150UL
10095-990EA 462.47 USD
10095-990
Anti-tim Rabbit Polyclonal Antibody
Antibodies
TIMELESS1 (TIM1) gene encodes protein timeless homolog belonging to timeless family. TIM1 is localized in nucleus and regulated by cell cycle. It expresses high levels in S, G2 and M phases, and low in G0 and G1 phases. TIM1 is involved in the circadian rhythm autoregulatory loop and is a crucial component of the circadian core oscillator, which includes the CRY proteins, CLOCK, NPAS2, etc. It is also involved in cell survival after DNA damage or replication stress and reported to play important role in epithelial cell morphogenesis and formation of branching tubules.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human spleen, 1:20-1:200; IP: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: tim
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR