Anti-TM Rabbit Polyclonal Antibody

Supplier: Proteintech
14318-1-AP-150UL
10095-958EA 462.47 USD
10095-958
Anti-TM Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: TM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR