Anti-TRX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13089-1-AP-150UL
10095-940EA 462.47 USD
10095-940
Anti-TRX2 Rabbit Polyclonal Antibody
Antibodies
TXN2(Thioredoxin-2) is also named as TRX2,MTRX,Mt-TRX and regulates the mitochondrial redox state and plays an important role in cell proliferation.It has been shown that cells conditionally deficient in Trx2 undergo apoptosis in the absence of exogenous stress, accompanied by an accumulation of intracellular ROS, the activation of caspase 9 and caspase 3, and the release of cytochrome c into the cytosol.In the retina, the expression profile and the role of Trx2 have not yet been defined.

Western Blot: Raji Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TRX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR