Anti-UAP1L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
25262-1-AP-150UL
10095-932EA 462.47 USD
10095-932
Anti-UAP1L1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MDA-MB-453s Cells, 1:200-1:2000

Type: Primary
Antigen: UAP1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR