Anti-TFPT Rabbit Polyclonal Antibody

Supplier: Proteintech
10097-2-AP-150UL
10095-846EA 462.47 USD
10095-846
Anti-TFPT Rabbit Polyclonal Antibody
Antibodies
TFPT/FB1 is one of three fusion partners of the E2A gene. TFPT/FB1 is expressed at low level in a variety of human tissues, including hemopoietic cell lines from different lineages. The function of the putative TFPT/FB1 protein, however, is currently unknown as database sequence comparisons have failed to reveal strong homology with known proteins. The E2A-TFPT/FB1 fusion appears to be a recurrent feature of pre-B ALLs, suggesting that it might have a role in the development and/or progression of leukemogenesis. The gene encoding TFPT/FB1 is localized on 19q13.4.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: TFPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR