Anti-Transferrin Mouse Monoclonal Antibody [clone: 2B4C6]

Supplier: Proteintech
66171-1-IG-150UL
10095-812EA 462.47 USD
10095-812
Anti-Transferrin Mouse Monoclonal Antibody [clone: 2B4C6]
Transferrin(TF) is also named as Beta-1 metal-binding globulin, Siderophilin and belongs to the transferrin family. It is a bilobal iron-binding protein that plays the crucial role of binding ferric iron and keeping it in solution, thereby controlling the levels of this important metal. Serum transferrin may have a further role in stimulating cell proliferation.

Western Blot: Human plasma Tissue, 1:1000-1:10000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: Transferrin
Clonality: Monoclonal
Clone: 2B4C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR