Anti-TCP11 Rabbit Polyclonal Antibody

Supplier: Proteintech
14606-1-AP-150UL
10095-746EA 462.47 USD
10095-746
Anti-TCP11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Testis Tissue, 1:1000-1:10000

Type: Primary
Antigen: TCP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR