Anti-TAB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14410-1-AP-150UL
10095-568EA 462.47 USD
10095-568
Anti-TAB2 Rabbit Polyclonal Antibody
Antibodies
Tab2 has a role in the inflammatory signal transduction pathway and is involved in the reversal of NCoR-dependent and NFkB- or APP-mediated gene repression and androgen antagonist response. Besides, it is identified as a cofactor for ERBB4 to assemble NCoR corepressor complex in response to neuregulin . It also functions as an adaptor that links TAK1 and TRAF6 in response to interleukin-1β (IL-1β) or tumour necrosis factor-α (TNF-α), and thereby mediates TAK1 activation .

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human malignant melanoma Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: TAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR