Anti-SCGF Rabbit Polyclonal Antibody

Supplier: Proteintech
55019-1-AP-150UL
10094-192EA 462.47 USD
10094-192
Anti-SCGF Rabbit Polyclonal Antibody
Antibodies
CLEC11A, also named as CLECSF3, LSLCL, SCGF and p47, stimulates the proliferation and differentiation of hematopoietic precursor cells from various lineages, including erythrocytes, lymphocytes, granulocytes and macrophages. CLEC11A acts synergistically with other cytokines, including IL-3, GCSF, GMCSF and FLT3 ligand. It suppresses SCF-stimulated erythrocyte proliferation. The antibody is specific to CLEC11A.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: SCGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR