Anti-SCC4 Rabbit Polyclonal Antibody

Supplier: Proteintech
24770-1-AP-150UL
10094-172EA 462.47 USD
10094-172
Anti-SCC4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: SCC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR