Anti-SACM1L Rabbit Polyclonal Antibody

Supplier: Proteintech
13033-1-AP-150UL
10094-110EA 462.47 USD
10094-110
Anti-SACM1L Rabbit Polyclonal Antibody
Antibodies
SACM1L(Suppressor of actin mutations 1-like protein) is also named as KIAA0851.It is a phosphoinositide phosphatase that hydrolyzes PtdIns3P and PtdIns4P and has low activity towards PtdIns(3,5)P2.

Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: SACM1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR