Anti-S100 A8 Rabbit Polyclonal Antibody

Supplier: Proteintech
15792-1-AP-150UL
10094-094EA 462.47 USD
10094-094
Anti-S100 A8 Rabbit Polyclonal Antibody
Antibodies
S100A8 may form homodimer or heterodimer with S100A9(16216873);

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human Stomach cancer Tissue, 1:20-1:200

Type: Primary
Antigen: S100 A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR