Anti-S100 A6 Rabbit Polyclonal Antibody

Supplier: Proteintech
10245-1-AP-150UL
10094-092EA 462.47 USD
10094-092
Anti-S100 A6 Rabbit Polyclonal Antibody
Antibodies
S100A6 is also named as calcyclin, prolactin receptor-associated protein (PRA), growth factor-inducible protein 2A9 ir MLN4. It belongs to S100 family of low molecular weight, acidic, calcium-binding proteins which contain two EF-hand calcium binding sites. S100A6 may function as a calcium sensor to activate several processes in the calcium signal transduction pathway of cell growth, proliferation, secretion and exocytosis. The antibody is specific for S100A6.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: S100 A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR