Anti-S100 A13 Rabbit Polyclonal Antibody

Supplier: Proteintech
14987-1-AP-150UL
10094-078EA 462.47 USD
10094-078
Anti-S100 A13 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF-7 Cells, 1:200-1:1000; IHC: Human thyroid cancer Tissue, 1:20-1:200

Type: Primary
Antigen: S100 A13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR