Anti-S1 tag Mouse Monoclonal Antibody [clone: 1A9B5]

Supplier: Proteintech
66165-1-IG-150UL
10094-068EA 462.47 USD
10094-068
Anti-S1 tag Mouse Monoclonal Antibody [clone: 1A9B5]
Antibodies
S1 tag, with sequence NANNPDWDF, is derived from the hepatitis B virus preS1 region. S1 tag is widely used in various protein expression systems. This antibody can detect the over-expressed fusion proteins containing S1 epitope tag.

Western Blot: Recombinant Protein, 1:5000-1:50000

Type: Primary
Antigen: S1 tag
Clonality: Monoclonal
Clone: 1A9B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR