Anti-RRP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14896-1-AP-150UL
10094-006EA 462.47 USD
10094-006
Anti-RRP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:1000-1:10000

Type: Primary
Antigen: RRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR