Anti-RPS7 Rabbit Polyclonal Antibody

Supplier: Proteintech
14491-1-AP-150UL
10093-984EA 462.47 USD
10093-984
Anti-RPS7 Rabbit Polyclonal Antibody
Antibodies
Ribosomal protein S7(RPS7) belongs to the RPs, which is structural compoent of the ribosome invilved in protein synthesis. It interacts with and inhibit mdm2 mediated p53 degradation function. Also RPS7 has a essential role in early development. RPS7 involves in the maturation of ribosomal RNAs in the large or the small ribosomal subunit production pathway

Western Blot:NIH/3T3 Cells, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: RPS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR