Anti-RPS4X Rabbit Polyclonal Antibody

Supplier: Proteintech
14799-1-AP-150UL
10093-972EA 462.47 USD
10093-972
Anti-RPS4X Rabbit Polyclonal Antibody
Antibodies
RPS4X, a X-linked gene in the human, encodes a subunit of ribosomal protein S4, a component of the 40S subunit and 60S subunit. Ribosomal protein S4 is the only ribosomal protein known to be encoded by more than one gene, namely this gene, RPS4Y2 and the ribosomal protein S4, X-linked (RPS4X).

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: RPS4X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR