Anti-RPS27L Rabbit Polyclonal Antibody

Supplier: Proteintech
15871-1-AP-150UL
10093-962EA 462.47 USD
10093-962
Anti-RPS27L Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:500-1:5000

Type: Primary
Antigen: RPS27L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR