Anti-RPS16 Rabbit Polyclonal Antibody

Supplier: Proteintech
15603-1-AP-150UL
10093-946EA 462.47 USD
10093-946
Anti-RPS16 Rabbit Polyclonal Antibody
Antibodies
RPS16, the 40S ribosmal protein S16, belongs to the ribosomal protein S9P family. It is located in the cytoplasm and can be acetylated. RPS16 is extensively involved in rRNA processing, translational elongation, initiation and termination via its RNA binding activity. Catalog#15603-1-AP is a rabbit polyclonal antibody raised against full-length RPS16 of human origin.

  • Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200; IF: MCF-7, 1:10-1:100

Type: Primary
Antigen: RPS16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR