Anti-RIPK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15366-1-AP-150UL
10093-700EA 462.47 USD
10093-700
Anti-RIPK2 Rabbit Polyclonal Antibody
Antibodies
RIPK2 is also named as CARDIAK, RICK, RIP2 and belongs to the receptor-interacting serine/threonine-protein kinase(RIPK) family. RIPK2 functions as a key signaling protein in host defense responses induced by activation of the cytosolic pattern recognition receptors (PRR) NOD1 and NOD2. It has also been reported to mediate other cellular responses, including TLR signaling, caspase 1 activation , apoptosis and autophagy. It has 2 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: RIPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR