Anti-RIC8B Rabbit Polyclonal Antibody

Supplier: Proteintech
17790-1-AP-150UL
10093-670EA 462.47 USD
10093-670
Anti-RIC8B Rabbit Polyclonal Antibody
Antibodies
Ric8, also known as synembryn, is a guanine nucleotide exchange factor (GEF) that enhances variegated G protein signaling pathways, including Gα(q) and Gα(s) regulation of neurotransmission, Gα(i)-dependent mitotic spindle positioning during (asymmetric) cell division, and Gα(olf)-dependent odorant receptor signaling . In mammals, two Ric8 homologues have been reported (Ric8A and Ric8B). Ric8B is able to potentiate G(s)/G(olf) signaling .

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: RIC8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR