Anti-PLOD3 Mouse Monoclonal Antibody [clone: 2G12B8]

Supplier: Proteintech
60058-1-IG-150UL
10092-482EA 462.47 USD
10092-482
Anti-PLOD3 Mouse Monoclonal Antibody [clone: 2G12B8]
Antibodies
PLOD3, also named as LH3, forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links. The major function of PLOD3 in osteoblasts is to glucosylate galactosylhydroxylysine residues in type I collagen.

Western Blot: Human Placenta, 1:200-1:1000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: PLOD3
Clonality: Monoclonal
Clone: 2G12B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR