Anti-PIP4K2B Rabbit Polyclonal Antibody

Supplier: Proteintech
13218-1-AP-150UL
10092-346EA 462.47 USD
10092-346
Anti-PIP4K2B Rabbit Polyclonal Antibody
Antibodies
PIP4K2B, also named as PIP5K2B, participates in the biosynthesis of phosphatidylinositol-4,5-bisphosphate. PIP4K2B has a homodimer. It binds TNFRSF1A(24/32/50kd).

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PIP4K2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR