Anti-PILRA Rabbit Polyclonal Antibody

Supplier: Proteintech
11818-1-AP-150UL
10092-278EA 462.47 USD
10092-278
Anti-PILRA Rabbit Polyclonal Antibody
Antibodies
Paired receptors consist of highly related activating and inhibitory receptors and are widely involved in the regulation of the immune system. Paired immunoglobulin-like type 2 receptor alpha (PILRα) and PILRβ are paired receptors bearing highly similar extracellular ligand-binding domains but divergent intracellular signaling domains. PILRα is the immune inhibitory receptor possessing an immunoreceptor tyrosine-based inhibitory motif (ITIM) in its cytoplasmic domain enabling it to deliver inhibitory signals. Binding of PILRα to its ligand CD99 is involved in immune regulation. PILRα can also recognize other ligands, PILR-associating neural protein (PANP), and HSV-1 glycoprotein B (HSV-1 gB).

Type: Primary
Antigen: PILRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR