Anti-PI16 Rabbit Polyclonal Antibody

Supplier: Proteintech
12267-1-AP-150UL
10092-222EA 462.47 USD
10092-222
Anti-PI16 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:200-1:1000

Type: Primary
Antigen: PI16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR