Anti-PHF19 Rabbit Polyclonal Antibody

Supplier: Proteintech
11895-1-AP-150UL
10092-190EA 462.47 USD
10092-190
Anti-PHF19 Rabbit Polyclonal Antibody
Antibodies
PHD FINGER PROTEIN 19(PHF19), a protein homologous to the Drosophila 'Polycomblike' (Pcl) protein, encoded by human gene PCL, which encodes 2 deduced proteins: a short form of 207 aa and a long form of 580 aa. The PCL family proteins contains a PCL homology domain, a Tudor domain, 2 PHD fingers and a C-terminal tail. It is highly expressed in thymus and heart, while 508aa form predominant in placenta, skeletal muscle and kidney and 207aa form in liver and peripheral blood leukocytes. Both of these forms can inhibit transcriptional activity. And the long form is corelated with cancer.Acts as a transcritpional repressor. The mRNA level of PHF19 is low in many normal tissue, it's elevated in cancer-derived cell lines.

Western Blot:rat Brain Tissue, 1:200-1:1000; IP: HeLa Cells, 1:200-1:2000; IF: Mouse ES Cells, 1:20-1:200

Type: Primary
Antigen: PHF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR