Anti-PDRG1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16968-1-AP-150UL
10092-052EA 462.47 USD
10092-052
Anti-PDRG1 Rabbit Polyclonal Antibody
Antibodies
PDRG1 codes for a novel protein called p53 and DNA damage-regulated protein 1. Human PDRG1 is predominantly expressed in normal testis and its expression can be induced by UV irradiation and be repressed by p53 . Recently, PDRG1 was identified as a novel tumor marker for multiple malignancies that is selectively regulated by genotoxic stress. Its expression was upregulated in multiple malignancies including cancers of the colon, rectum, ovary, lung, stomach, breast and uterus when compared to normal tissues , suggesting its being a novel tumor marker that could play a role in cancer development and/or progression.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PDRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR