Anti-PCDH11X Rabbit Polyclonal Antibody

Supplier: Proteintech
20070-1-AP-150UL
10091-916EA 462.47 USD
10091-916
Anti-PCDH11X Rabbit Polyclonal Antibody
Antibodies
PCDH11X, also named as KIAA1326, PCDH11 and PCDHX, is a potential calcium-dependent cell-adhesion protein. The antibody recognizes isoforms1 and isoform6 of PCDH11X. The antibody has no cross reaction to PCDH11Y.

Western Blot: Human Brain Tissue, 1:200-1:1000; IHC: Human Brain, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PCDH11X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR