Anti-OTOP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24456-1-AP-150UL
10091-632EA 462.47 USD
10091-632
Anti-OTOP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:200-1:1000

Type: Primary
Antigen: OTOP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR